Assay Details
| Gene Symbol: MRPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285279.1 | NP_001272208.1 | 4 | 823 | 91 | |
| XM_005551511.2 | XP_005551568.1 | 4 | 1362 | 91 | ||
| GenBank mRNA | AB171133.1 | - | 1 | 814 | 91 | |
| AB172555.1 | - | 4 | 824 | 91 | ||
More Information
Set Membership: |
|

MRPL18
Genomic Map