Assay Details
| Gene Symbol: DNAJA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284713.1 | NP_001271642.1 | 9 | 1221 | 129 | |
| GenBank mRNA | AB169031.1 | - | 9 | 1221 | 129 | |
More Information
Set Membership: |
|

DNAJA1
Genomic Map