Assay Details
| Gene Symbol: RARRES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546230.2 | XP_005546287.1 | 1 - 2 | 542 | 113 | |
| XM_015445242.1 | XP_015300728.1 | 1 - 2 | 542 | 113 | ||
More Information
Set Membership: |
|

RARRES1
Genomic Map