Assay Details
| Gene Symbol: RPL22L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546366.2 | XP_005546423.1 | 3 - 4 | 323 | 115 | |
| XM_005546367.2 | XP_005546424.1 | 3 - 4 | 530 | 115 | ||
More Information
Set Membership: |
|

RPL22L1
Genomic Map