Assay Details
| Gene Symbol: PLCXD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548191.2 | XP_005548248.1 | 1 - 2 | 748 | 85 | |
| XM_015446457.1 | XP_015301943.1 | 1 - 2 | 748 | 85 | ||
| XM_015446458.1 | XP_015301944.1 | 1 - 2 | 748 | 85 | ||
| XM_015446459.1 | XP_015301945.1 | 1 - 2 | 748 | 85 | ||
| XM_015446460.1 | XP_015301946.1 | 1 - 2 | 748 | 85 | ||
| XM_015446461.1 | XP_015301947.1 | 1 - 2 | 748 | 85 | ||
More Information
Set Membership: |
|

PLCXD2
Genomic Map