Assay Details
Gene Symbol: RBM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550235.2 | XP_005550292.1 | 2 - 3 | 448 | 106 | |
XM_005550236.2 | XP_005550293.1 | 2 - 3 | 353 | 106 | ||
XM_005550237.2 | XP_005550294.1 | 2 - 3 | 568 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
