Assay Details
| Gene Symbol: BMF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559153.2 | XP_005559210.1 | 5 - 6 | 796 | 65 | |
| XM_015452634.1 | XP_015308120.1 | 4 - 5 | 1062 | 65 | ||
| XM_015452635.1 | XP_015308121.1 | 4 - 5 | 715 | 65 | ||
| XM_015452636.1 | XP_015308122.1 | 3 - 4 | 1775 | 65 | ||
| XM_015452637.1 | XP_015308123.1 | 3 - 4 | 1251 | 65 | ||
| XM_015452638.1 | XP_015308124.1 | 3 - 4 | 1679 | 65 | ||
More Information
Set Membership: |
|

BMF
Genomic Map