Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567628.2 | XP_005567685.1 | 1 - 2 | 1458 | 101 | |
| XM_015457518.1 | XP_015313004.1 | 3 - 4 | 2757 | 101 | ||
More Information
Set Membership: |
|

LIF
Genomic Map