Assay Details
| Gene Symbol: IL18R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575164.2 | XP_005575221.1 | 11 - 12 | 1566 | 114 | |
| XM_005575165.2 | XP_005575222.1 | 11 - 12 | 1560 | 114 | ||
| XM_015433326.1 | XP_015288812.1 | 10 - 11 | 1773 | 114 | ||
More Information
Set Membership: |
|

IL18R1
Genomic Map