Assay Details
| Gene Symbol: LOC102141352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581620.2 | XP_005581677.1 | 1 | 347 | 74 | |
| Gene Symbol: LOC102141744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581621.2 | XP_005581678.2 | 1 | 347 | 74 | |
More Information
Set Membership: |
|

LOC102141352
Genomic Map