Assay Details
| Gene Symbol: LOC102126770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594666.2 | XP_005594723.1 | 10 - 11 | 1514 | 100 | |
| XM_005594669.2 | XP_005594726.1 | 10 - 11 | 1500 | 100 | ||
| XM_005594671.2 | XP_005594728.1 | 10 - 11 | 1359 | 100 | ||
More Information
Set Membership: |
|

LOC102126770
Genomic Map