Assay Details
| Gene Symbol: LOC102126770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594666.2 | XP_005594723.1 | 12 - 13 | 1975 | 100 | |
| XM_005594667.2 | XP_005594724.1 | 12 - 13 | 1970 | 100 | ||
| XM_005594669.2 | XP_005594726.1 | 12 - 13 | 1961 | 100 | ||
| XM_005594670.2 | XP_005594727.1 | 11 - 12 | 1835 | 100 | ||
| XM_005594671.2 | XP_005594728.1 | 12 - 13 | 1820 | 100 | ||
More Information
Set Membership: |
|

LOC102126770
Genomic Map