Assay Details
| Gene Symbol: LOC102126770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594666.2 | XP_005594723.1 | 7 - 8 | 1093 | 61 | |
| XM_005594667.2 | XP_005594724.1 | 7 - 8 | 1094 | 61 | ||
| XM_005594669.2 | XP_005594726.1 | 7 - 8 | 1079 | 61 | ||
| XM_005594670.2 | XP_005594727.1 | 7 - 8 | 1094 | 61 | ||
| XM_005594671.2 | XP_005594728.1 | 7 - 8 | 938 | 61 | ||
More Information
Set Membership: |
|

LOC102126770
Genomic Map