Assay Details
Gene Symbol: FCAMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449247.1 | XP_015304733.1 | 3 - 4 | 668 | 105 | |
XM_015449249.1 | XP_015304735.1 | 2 - 3 | 751 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
