Assay Details
| Gene Symbol: GCSAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548175.2 | XP_005548232.1 | 5 - 6 | 408 | 99 | |
| XM_005548176.2 | XP_005548233.1 | 5 - 6 | 451 | 99 | ||
More Information
Set Membership: |
|

GCSAM
Genomic Map