Assay Details
| Gene Symbol: PRSS42P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015445791.1 | XP_015301277.1 | 4 - 5 | 2801 | 90 | |
| XM_015445792.1 | XP_015301278.1 | 4 - 5 | 2800 | 90 | ||
More Information
Set Membership: |
|

PRSS42P
Genomic Map