Assay Details
| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554647.2 | XP_005554704.1 | 5 - 6 | 6782 | 74 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

NWD2| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554647.2 | XP_005554704.1 | 5 - 6 | 6782 | 74 | |