Assay Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557498.2 | XP_005557555.1 | 4 - 5 | 1768 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005557498.2 | XP_005557555.1 | 4 - 5 | 1768 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |