Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559617.2 | XP_005559674.2 | 1 - 2 | 384 | 95 | |
| XM_015452858.1 | XP_015308344.1 | 1 - 2 | 384 | 95 | ||
| XM_015452859.1 | XP_015308345.1 | 1 - 2 | 385 | 95 | ||
| XM_015452860.1 | XP_015308346.1 | 1 - 2 | 383 | 95 | ||
| XM_015452861.1 | XP_015308347.1 | 1 - 2 | 383 | 95 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map