Assay Details
| Gene Symbol: SUV39H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319605.1 | NP_001306534.1 | 3 - 4 | 873 | 91 | |
| XM_005564674.2 | XP_005564731.1 | 3 - 4 | 1169 | 91 | ||
| XM_005564675.2 | XP_005564732.1 | 3 - 4 | 970 | 91 | ||
| XM_005564676.2 | XP_005564733.1 | 3 - 4 | 840 | 91 | ||
| XM_005564677.2 | XP_005564734.1 | 3 - 4 | 884 | 91 | ||
| XM_005564678.2 | XP_005564735.1 | 2 - 3 | 774 | 91 | ||
| XM_005564680.2 | XP_005564737.1 | 2 - 3 | 812 | 91 | ||
| XM_005564681.2 | XP_005564738.1 | 2 - 3 | 692 | 91 | ||
| XM_015455588.1 | XP_015311074.1 | 3 - 4 | 1154 | 91 | ||
| GenBank mRNA | AB179326.1 | - | 3 - 4 | 873 | 91 | |
More Information
Set Membership: |
|

SUV39H2
Genomic Map