Assay Details
| Gene Symbol: RDH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571270.2 | XP_005571327.1 | 3 - 4 | 2448 | 81 | |
| XM_015431070.1 | XP_015286556.1 | 3 - 4 | 1817 | 81 | ||
More Information
Set Membership: |
|

RDH16
Genomic Map