Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577191.2 | XP_005577248.1 | 2 - 3 | 435 | 94 | |
| XM_015434429.1 | XP_015289915.1 | 2 - 3 | 1191 | 94 | ||
More Information
Set Membership: |
|

GAL3ST3
Genomic Map