Assay Details
| Gene Symbol: LOC102140322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584300.2 | XP_005584357.1 | 4 - 5 | 2107 | 86 | |
| XM_015438527.1 | XP_015294013.1 | 2 - 3 | 2328 | 86 | ||
More Information
Set Membership: |
|

LOC102140322
Genomic Map