Assay Details
| Gene Symbol: ZNF627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588067.2 | XP_005588124.1 | 3 - 4 | 397 | 76 | |
| XM_015440566.1 | XP_015296052.1 | 3 - 4 | 397 | 76 | ||
| XM_015440567.1 | XP_015296053.1 | 3 - 4 | 397 | 76 | ||
More Information
Set Membership: |
|

ZNF627
Genomic Map