Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540997.2 | XP_005541054.1 | 1 - 2 | 220 | 84 | |
| GenBank mRNA | AB169197.1 | - | 1 - 2 | 164 | 84 | |
More Information
Set Membership: |
|

SRP9
Genomic Map