Assay Details
| Gene Symbol: PIGZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015444872.1 | XP_015300358.1 | 2 - 3 | 1494 | 68 | |
| XM_015444873.1 | XP_015300359.1 | 3 - 4 | 1307 | 68 | ||
| XM_015444874.1 | XP_015300360.1 | 3 - 4 | 486 | 68 | ||
More Information
Set Membership: |
|

PIGZ
Genomic Map