Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553726.2 | XP_005553783.1 | 2 - 3 | 1789 | 77 | |
| XM_005553727.2 | XP_005553784.1 | 2 - 3 | 1769 | 77 | ||
| XM_015449706.1 | XP_015305192.1 | 1 - 2 | 3624 | 77 | ||

PGBD1
Genomic Map