Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553726.2 | XP_005553783.1 | 3 - 4 | 1942 | 101 | |
| XM_005553727.2 | XP_005553784.1 | 3 - 4 | 1922 | 101 | ||
| XM_015449706.1 | XP_015305192.1 | 2 - 3 | 3777 | 101 | ||
More Information
Set Membership: |
|

PGBD1
Genomic Map