Assay Details
| Gene Symbol: SLC25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562203.2 | XP_005562260.1 | 3 - 4 | 481 | 60 | |
| XM_005562204.2 | XP_005562261.1 | 4 - 5 | 553 | 60 | ||
| XM_015454205.1 | XP_015309691.1 | 3 - 4 | 4349 | 60 | ||
| XM_015454206.1 | XP_015309692.1 | 4 - 5 | 2017 | 60 | ||
More Information
Set Membership: |
|

SLC25A29
Genomic Map