Assay Details
| Gene Symbol: FNTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563241.2 | XP_005563298.1 | 4 - 5 | 1136 | 84 | |
| GenBank mRNA | AB171159.1 | - | 4 - 5 | 523 | 84 | |
More Information
Set Membership: |
|

FNTA
Genomic Map