Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567278.2 | XP_005567335.1 | 3 - 4 | 348 | 109 | |
| XM_005567280.2 | XP_005567337.1 | 4 - 5 | 472 | 109 | ||
| XM_005567281.2 | XP_005567338.1 | 3 - 4 | 269 | 109 | ||
| XM_015457200.1 | XP_015312686.1 | 5 - 6 | 781 | 109 | ||
| XM_015457201.1 | XP_015312687.1 | 5 - 6 | 781 | 109 | ||
| XM_015457202.1 | XP_015312688.1 | 4 - 5 | 478 | 109 | ||
| XM_015457203.1 | XP_015312689.1 | 4 - 5 | 420 | 109 | ||
| XM_015457204.1 | XP_015312690.1 | 4 - 5 | 382 | 109 | ||
| XM_015457208.1 | XP_015312694.1 | 4 - 5 | 606 | 109 | ||
| XM_015457209.1 | XP_015312695.1 | 4 - 5 | 578 | 109 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map