Assay Details
| Gene Symbol: ATF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589977.2 | XP_005590034.1 | 2 - 3 | 1164 | 80 | |
| XM_005589978.2 | XP_005590035.1 | 2 - 3 | 528 | 80 | ||
More Information
Set Membership: |
|

ATF5
Genomic Map