Assay Details
| Gene Symbol: TMEM133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596039.2 | XP_005596096.1 | 1 | 332 | 83 | |
| Gene Symbol: ARHGAP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579430.2 | XP_005579487.1 | 24 | 6966 | 83 | |
More Information
Set Membership: |
|

TMEM133
Genomic Map