Assay Details
| Gene Symbol: LOC102127568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544726.2 | XP_005544783.1 | 6 - 7 | 725 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102127568| Gene Symbol: LOC102127568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544726.2 | XP_005544783.1 | 6 - 7 | 725 | 95 | |
Set Membership: |
|