Assay Details
| Gene Symbol: LOC102146742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546419.2 | XP_005546476.1 | 8 - 9 | 2207 | 100 | |
| XM_015445537.1 | XP_015301023.1 | 7 - 8 | 2063 | 100 | ||
More Information
Set Membership: |
|

LOC102146742
Genomic Map