Assay Details
| Gene Symbol: LOC102115705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546925.2 | XP_005546982.1 | 1 - 2 | 362 | 69 | |
| XM_005546926.2 | XP_005546983.1 | 1 - 2 | 366 | 69 | ||
More Information
Set Membership: |
|

LOC102115705
Genomic Map