Assay Details
| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015446565.1 | XP_015302051.1 | 7 - 8 | 1489 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABRR3| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015446565.1 | XP_015302051.1 | 7 - 8 | 1489 | 83 | |
Set Membership: |
|