Assay Details
Gene Symbol: LOC102128285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015448507.1 | XP_015303993.1 | 2 | 3434 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102128285 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015448507.1 | XP_015303993.1 | 2 | 3434 | 114 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |