Assay Details
| Gene Symbol: C6H5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557806.2 | XP_005557863.1 | 1 - 2 | 256 | 96 | |
| XM_005557809.2 | XP_005557866.1 | 1 - 2 | 256 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005557806.2, XM_005557809.2 transcripts.
Set Membership: |
|

C6H5orf24
Genomic Map