Assay Details
Gene Symbol: TNFAIP8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559553.2 | XP_005559610.1 | 1 - 2 | 364 | 96 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TNFAIP8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005559553.2 | XP_005559610.1 | 1 - 2 | 364 | 96 |