Assay Details
| Gene Symbol: SNX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015429889.1 | XP_015285375.1 | 1 - 2 | 875 | 84 | |
| XM_015429890.1 | XP_015285376.1 | 1 - 2 | 875 | 84 | ||
More Information
Set Membership: |
|

SNX21
Genomic Map