Assay Details
| Gene Symbol: SLC2A4RG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569613.2 | XP_005569670.1 | 5 - 6 | 1504 | 81 | |
| XM_005569614.1 | XP_005569671.1 | 5 - 6 | 1501 | 81 | ||
More Information
Set Membership: |
|

SLC2A4RG
Genomic Map