Assay Details
| Gene Symbol: LOC102120532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579495.2 | XP_005579552.1 | 6 - 7 | 1027 | 67 | |
| XM_015435645.1 | XP_015291131.1 | 5 - 6 | 1558 | 67 | ||
| XM_015435646.1 | XP_015291132.1 | 5 - 6 | 1556 | 67 | ||
More Information
Set Membership: |
|

LOC102120532
Genomic Map