Assay Details
Gene Symbol: NT5M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583019.2 | XP_005583076.1 | 1 - 2 | 420 | 78 | |
XM_005583020.2 | XP_005583077.1 | 1 - 2 | 420 | 78 | ||
XM_005583021.2 | XP_005583078.1 | 1 - 2 | 462 | 78 | ||
XM_015437714.1 | XP_015293200.1 | 1 - 2 | 420 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
