Assay Details
| Gene Symbol: LOC102132489 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015440682.1 | XP_015296168.1 | 7 - 8 | 1395 | 100 | |
| XM_015440683.1 | XP_015296169.1 | 7 - 8 | 1336 | 100 | ||
| XM_015440684.1 | XP_015296170.1 | 7 - 8 | 1447 | 100 | ||
More Information
Set Membership: |
|

LOC102132489
Genomic Map