Assay Details
| Gene Symbol: MRPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588374.2 | XP_005588431.1 | 1 - 2 | 563 | 76 | |
| GenBank mRNA | AB179449.1 | - | 1 - 2 | 103 | 76 | |
More Information
Set Membership: |
|

MRPL34
Genomic Map