Assay Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590152.2 | XP_005590209.2 | 4 - 5 | 1528 | 90 | |
| XM_005590154.2 | XP_005590211.1 | 6 - 7 | 705 | 90 | ||
| XM_015441339.1 | XP_015296825.1 | 7 - 8 | 820 | 90 | ||
| XM_015441340.1 | XP_015296826.1 | 6 - 7 | 650 | 90 | ||
More Information
Set Membership: |
|

ZNF577
Genomic Map