Assay Details
| Gene Symbol: OGFOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591967.2 | XP_005592024.1 | 2 - 3 | 409 | 93 | |
| GenBank mRNA | AB173727.1 | - | 2 - 3 | 353 | 93 | |
More Information
Set Membership: |
|

OGFOD1
Genomic Map