Assay Details
| Gene Symbol: BEX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594227.2 | XP_005594284.1 | 2 - 3 | 350 | 95 | |
| XM_015443809.1 | XP_015299295.1 | 1 - 2 | 85 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005594227.2, XM_015443809.1 transcripts.
Set Membership: |
|

BEX3
Genomic Map