Assay Details
| Gene Symbol: HHLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548228.2 | XP_005548285.1 | 5 - 6 | 1163 | 79 | |
| XM_015446481.1 | XP_015301967.1 | 6 - 7 | 1241 | 79 | ||
| XM_015446482.1 | XP_015301968.1 | 5 - 6 | 2198 | 79 | ||
| XM_015446483.1 | XP_015301969.1 | 5 - 6 | 975 | 79 | ||
More Information
Set Membership: |
|

HHLA2
Genomic Map